3-(Methoxydimethylsilyl)propyl acrylate ≥95.0% (by GC) stabilized
Supplier: TCI America
Synonyms:
Acrylic Acid 3-(Methoxydimethylsilyl)propyl Ester
M2525-5G
TCM2525-5GEA
249
USD
TCM2525-5G
3-(Methoxydimethylsilyl)propyl acrylate ≥95.0% (by GC) stabilized
3-(Methoxydimethylsilyl)propyl acrylate
(stabilized with MEHQ)
CAS Number: 111918-90-2
MDL Number: MFCD00054801
Molecular Formula: C9H18O3Si
Molecular Weight: 202.33
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.95
Storage Temperature:
Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: Acrylic Acid 3-(Methoxydimethylsilyl)propyl Ester
Formula:
C₉H₁₈O₃Si MW: 202.33 g/mol Density: 0.95 (20 °C) |
MDL Number:
MFCD00054801 CAS Number: 111918-90-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...