β-Methyl-γ-octanolactone (mixture of isomers) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-Butyl-4-methyldihydro-2(3H)-furanone
M2516-25ML
TCM2516-25MLEA
95.36
USD
TCM2516-25ML
β-Methyl-γ-octanolactone (mixture of isomers) ≥98.0% (by GC)
beta-Methyl-gamma-octanolactone (mixture of isomers)
CAS Number: 39212-23-2
MDL Number: MFCD00209540
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 113
Specific Gravity (20/20): 0.96
Other Product Names / Synonyms: 5-Butyl-4-methyldihydro-2(3H)-furanone
Formula:
C₉H₁₆O₂ MW: 156.23 g/mol Boiling Pt: 64 °C Density: 0.96 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00209540 CAS Number: 39212-23-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...