Methyl-5-bromo-2-chloronicotinate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Methyl 5-bromo-2-chloronicotinate
CAS Number: 78686-79-0
MDL Number: MFCD03844848
Molecular Formula: C7H5BrClNO2
Molecular Weight: 250.48
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 52
Flash Point (°C): 110
Other Product Names / Synonyms: 5-Bromo-2-chloronicotinic Acid Methyl Ester, Methyl 5-Bromo-2-chloro-3-pyridinecarboxylate, 5-Bromo-2-chloro-3-pyridinecarboxylic Acid Methyl Ester
Formula:
C₇H₅BrClNO₂ MW: 250.48 g/mol Melting Pt: 52 °C Flash Pt: 110 °C |
MDL Number:
MFCD03844848 CAS Number: 78686-79-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...