2-Methyl-1,10-phenanthroline ≥98.0% (by HPLC)
Supplier: TCI America
M2512-1G
TCM2512-1GEA
272
USD
TCM2512-1G
2-Methyl-1,10-phenanthroline ≥98.0% (by HPLC)
2-Methyl-1,10-phenanthroline
CAS Number: 3002-77-5
MDL Number: MFCD06643855
Molecular Formula: C13H10N2
Molecular Weight: 194.24
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 80
Lambda max.: 267 nm (H2O (pH10))
Formula:
C₁₃H₁₀N₂ MW: 194.24 g/mol Boiling Pt: 175 °C Melting Pt: 80 °C |
MDL Number:
MFCD06643855 CAS Number: 3002-77-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...