Methyl cyclopentanecarboxylate ≥98.0% (by GC)
Supplier: TCI America
M2510-5G
TCM2510-5GEA
134
USD
TCM2510-5G
Methyl cyclopentanecarboxylate ≥98.0% (by GC)
Methyl cyclopentanecarboxylate
CAS Number: 4630-80-2
MDL Number: MFCD00019290
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: Cyclopentanecarboxylic Acid Methyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...