3-Cyano-6-methylchromone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 6-Methyl-4-oxo-4H-chromene-3-carbonitrile6-Methylchromone-3-carbonitrile

M2459-1G M2459-5G
TCM2459-1GEA 16.24 USD
TCM2459-1G TCM2459-5G
3-Cyano-6-methylchromone ≥98.0% (by HPLC, total nitrogen)
3-Cyano-6-methylchromone

CAS Number: 50743-18-5
MDL Number: MFCD00191962
Molecular Formula: C11H7NO2
Molecular Weight: 185.18
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: 3-Cyano-6-methylchromone, 6-Methyl-4-oxo-4H-1-benzopyran-3-carbonitrile

Formula: C₁₁H₇NO₂
MW: 185.19 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00191962
CAS Number: 50743-18-5
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/84/83/28988483.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR