7-Methoxy-1-naphthylacetonitrile ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 7-Methoxy-1-naphthylmethylcyanide

M2421-1G M2421-5G
TCM2421-1GEA 33.48 USD
TCM2421-1G TCM2421-5G
7-Methoxy-1-naphthylacetonitrile ≥99.0% (by GC)
7-Methoxy-1-naphthylacetonitrile

CAS Number: 138113-08-3
MDL Number: MFCD08704309
Molecular Formula: C13H11NO
Molecular Weight: 197.24
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 7-Methoxy-1-naphthylmethylcyanide

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Melting Pt: 84 °C
MDL Number: MFCD08704309
CAS Number: 138113-08-3
https://us.vwr.com/stibo/web/std.lang.all/84/52/28988452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR