2-Morpholino-8-phenylchromone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyranLY 294002

M2410-25MG
TCM2410-25MGEA 246.65 USD
TCM2410-25MG
2-Morpholino-8-phenylchromone ≥98.0% (by HPLC)
2-Morpholino-8-phenylchromone

CAS Number: 154447-36-6
MDL Number: MFCD00270881
Molecular Formula: C19H17NO3
Molecular Weight: 307.35
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 184
Storage Temperature: 0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran, LY 294002

Formula: C₁₉H₁₇NO₃
MW: 307.35 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00270881
CAS Number: 154447-36-6
https://us.vwr.com/stibo/web/std.lang.all/84/40/28988440.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR