4-Methoxypyridine-N-oxide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2401-5G
M2401-25G
TCM2401-5GEA
83.65
USD
TCM2401-5G
TCM2401-25G
4-Methoxypyridine-N-oxide ≥98.0% (by GC, titration analysis)
4-Methoxypyridine N-oxide
CAS Number: 1122-96-9
MDL Number: MFCD00006207
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 112
Lambda max.: 282 nm (CH3CN)
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD00006207 CAS Number: 1122-96-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...