3-(Methylthio)propyl acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Acetic Acid 3-(Methylthio)propyl Ester

M2386-25G
TCM2386-25GEA 149.16 USD
TCM2386-25G
3-(Methylthio)propyl acetate ≥98.0% (by GC)
3-(Methylthio)propyl acetate

CAS Number: 16630-55-0
MDL Number: MFCD00053113
Molecular Formula: C6H12O2S
Molecular Weight: 148.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Acetic Acid 3-(Methylthio)propyl Ester

Formula: C₆H₁₂O₂S
MW: 148.22 g/mol
Boiling Pt: 96 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00053113
CAS Number: 16630-55-0
https://us.vwr.com/stibo/web/std.lang.all/84/15/28988415.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR