Methyl-2-bromohexanoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2-bromohexanoate

M2378-250G M2378-25G
TCM2378-25GEA 18 USD
TCM2378-25G TCM2378-250G
Methyl-2-bromohexanoate ≥98.0% (by GC)
Methyl 2-bromohexanoate

CAS Number: 5445-19-2
MDL Number: MFCD00045038
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 69
Specific Gravity (20/20): 1.29


Other Product Names / Synonyms: 2-Bromohexanoic Acid Methyl Ester

https://us.vwr.com/stibo/web/std.lang.all/84/07/28988407.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR