Myoseverin ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9-Isopropyl-2,6-bis(4-methoxybenzylamino)purine
M2373-10MG
TCM2373-10MGEA
211
USD
TCM2373-10MG
Myoseverin ≥98.0% (by HPLC)
Myoseverin
CAS Number: 267402-71-1
MDL Number: MFCD02683596
Molecular Formula: C24H28N6O2
Molecular Weight: 432.53
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 108
Storage Temperature:
Product Categories: Bioscience, Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 9-Isopropyl-2,6-bis(4-methoxybenzylamino)purine
Formula:
C₂₄H₂₈N₆O₂ MW: 432.53 g/mol Melting Pt: 108 °C |
MDL Number:
MFCD02683596 CAS Number: 267402-71-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...