Myricitrin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone 3-O-Rhamnoside

M2361-50MG M2361-10MG
TCM2361-10MGEA 79.06 USD
TCM2361-10MG TCM2361-50MG
Myricitrin ≥98.0% (by HPLC)
Myricitrin

CAS Number: 17912-87-7
MDL Number: MFCD00016930
Molecular Formula: C21H20O12
Molecular Weight: 464.38
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 197
Specific rotation [a]20/D: -152 deg (C=0.5, MeOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Glycosides, Glycosides, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone 3-O-Rhamnoside

Formula: C₂₁H₂₀O₁₂
MW: 464.38 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00016930
CAS Number: 17912-87-7
https://us.vwr.com/stibo/web/std.lang.all/83/86/28988386.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR