2-Methyl-1,8-naphthyridine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Methyl-1,8-diazanaphthalene
, 2-Methyl[1,8]-Naphthyridine
, 2-Methylpyrido[2,3-b]pyridine
M2352-1G
TCM2352-1GEA
129
USD
TCM2352-1G
2-Methyl-1,8-naphthyridine ≥98.0% (by GC)
2-Methyl-1,8-naphthyridine
CAS Number: 1569-16-0
MDL Number: MFCD04066717
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 100
Other Product Names / Synonyms: 2-Methyl-1,8-diazanaphthalene, 2-Methylpyrido[2,3-b]pyridine
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD04066717 CAS Number: 1569-16-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...