Mitomycin C ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Ametycin
M2320-10MG
M2320-50MG
TCM2320-50MGEA
551.77
USD
TCM2320-50MG
TCM2320-10MG
Mitomycin C ≥98.0% (by HPLC)
Mitomycin C
CAS Number: 50-07-7
MDL Number: MFCD00078109
Molecular Formula: C15H18N4O5
Molecular Weight: 334.33
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 64
Storage Temperature: 0-10°C
Formula:
C₁₅H₁₈N₄O₅ MW: 334.33 g/mol Melting Pt: 64 °C |
MDL Number:
MFCD00078109 CAS Number: 50-07-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...