Methyl malonyl chloride ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 3-chloro-3-oxopropanoate

M2315-5G M2315-25G
TCM2315-25GEA 175 USD
TCM2315-25G TCM2315-5G
Methyl malonyl chloride ≥97.0% (by GC, titration analysis)
Methyl malonyl chloride

CAS Number: 37517-81-0
MDL Number: MFCD00013657
Molecular Formula: C4H5ClO3
Molecular Weight: 136.53
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 80
Specific Gravity (20/20): 1.28
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Methyl (Chloroformyl)acetate, (Chloroformyl)acetic Acid Methyl Ester, Methyl 3-Chloro-3-oxopropionate, 3-Chloro-3-oxopropionic Acid Methyl Ester

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 59 °C
Density: 1.28 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00013657
CAS Number: 37517-81-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/83/21/28988321.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR