(4-Methoxybenzyl)methyl ether ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Methoxymethyl)anisole
, 1-Methoxy-4-(methoxymethyl)benzene
M2305-5G
TCM2305-5GEA
214.07
USD
TCM2305-5G
(4-Methoxybenzyl)methyl ether ≥98.0% (by GC)
(4-Methoxybenzyl)methyl ether
CAS Number: 1515-81-7
MDL Number: MFCD00155135
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 226
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 4-(Methoxymethyl)anisole
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 226 °C Density: 1.04 (20 °C) |
MDL Number:
MFCD00155135 CAS Number: 1515-81-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...