2'-O-Methyladenosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol
M2291-1G
TCM2291-1GEA
427.32
USD
TCM2291-1G
2'-O-Methyladenosine ≥98.0% (by HPLC, titration analysis)
2'-O-Methyladenosine
CAS Number: 2140-79-6
MDL Number: MFCD00056002
Molecular Formula: C11H15N5O4
Molecular Weight: 281.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 208
Specific rotation [a]20/D: -57 deg (C=1, H2O)
Lambda max.: 259 nm (H2O)Storage Temperature:
Formula:
C₁₁H₁₅N₅O₄ MW: 281.27 g/mol Melting Pt: 208 °C |
MDL Number:
MFCD00056002 CAS Number: 2140-79-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...