Minocycline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M2288-1G M2288-5G
TCM2288-1GEA 173.49 USD
TCM2288-1G TCM2288-5G
Minocycline hydrochloride ≥98.0% (by HPLC, titration analysis)
Minocycline hydrochloride

CAS Number: 13614-98-7
MDL Number: MFCD00083669
Molecular Formula: C23H27N3O7
Molecular Weight: 493.94
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -175 deg (C=0.5, 0.1mL/L HCl)
Storage Temperature: 0-10°C

Formula: C₂₃H₂₇N₃O₇
MDL Number: MFCD00083669
CAS Number: 13614-98-7
https://us.vwr.com/stibo/web/std.lang.all/82/91/28988291.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR