S-Methyl thioacetate ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
S-Methylthioacetate
M2286-5G
M2286-25G
TCM2286-5GEA
78.45
USD
TCM2286-5G
TCM2286-25G
S-Methyl thioacetate ≥95.0% (by GC)
S-Methyl thioacetate
CAS Number: 1534-08-3
MDL Number: MFCD00014989
Molecular Formula: C3H6OS
Molecular Weight: 90.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 12
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: Thioacetic Acid S-Methyl Ester
Formula:
C₃H₆OS MW: 90.15 g/mol Boiling Pt: 98 °C Density: 1.03 (20 °C) Flash Pt: 12 °C |
MDL Number:
MFCD00014989 CAS Number: 1534-08-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...