Methyl propyl sulfide ≥98.0% (by GC)
Supplier: TCI America
M2285-25G
TCM2285-25GEA
75.09
USD
TCM2285-25G
Methyl propyl sulfide ≥98.0% (by GC)
Methyl propyl sulphide
CAS Number: 3877-15-4
MDL Number: MFCD00015219
Molecular Formula: C4H10S
Molecular Weight: 90.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 97
Melting point (°C): -113
Flash Point (°C): 11
Specific Gravity (20/20): 0.85
Formula:
C₄H₁₀S Boiling Pt: 97 °C Melting Pt: –113 °C Density: 0.85 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00015219 CAS Number: 3877-15-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...