Methyl-3-cyclopropyl-3-oxopropanoate ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-Cyclopropyl-3-oxopropionate3-Cyclopropyl-3-oxopropionic acid methyll esterMethyl 3-cyclopropyl-3-oxopropanoate

M2277-25G M2277-5G
TCM2277-25GEA 101.47 USD
TCM2277-25G TCM2277-5G
Methyl-3-cyclopropyl-3-oxopropanoate ≥96.0% (by GC)
Methyl-3-cyclopropyl-3-oxopropanoate

CAS Number: 32249-35-7
MDL Number: MFCD00274090
Molecular Formula: C7H10O3
Molecular Weight: 142.15
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 74
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 3-Cyclopropyl-3-oxopropionic Acid Methyl Ester

Formula: C7H10O3
MW: 142.1500 g/mol
Boiling Pt: 80 °C
Density: 1.10 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00274090
CAS Number: 32249-35-7
https://us.vwr.com/stibo/web/std.lang.all/82/80/28988280.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR