Mevastatin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Compactin

M2275-1G M2275-5G
TCM2275-1GEA 251.24 USD
TCM2275-1G TCM2275-5G
Mevastatin ≥98.0% (by HPLC)
Mevastatin

CAS Number: 73573-88-3
MDL Number: MFCD05662341
Molecular Formula: C23H34O5
Molecular Weight: 390.52
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 281 deg (C=0.4, Acetone)
Lambda max.: 246 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Antilipemic Ingredients, Prodrug Ingredients
Other Product Names / Synonyms: Compactin

Formula: C₂₃H₃₄O₅
MW: 390.52 g/mol
Melting Pt: 153 °C
MDL Number: MFCD05662341
CAS Number: 73573-88-3
https://us.vwr.com/stibo/web/std.lang.all/82/76/28988276.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR