Mevalonic lactone methacrylate ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: Methacrylic Acid 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Ester4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Methacrylate

M2261-1G
TCM2261-1GEA 431.11 USD
TCM2261-1G
Mevalonic lactone methacrylate ≥98.0% (by GC) stabilized
Mevalonic lactone methacrylate

(stabilized with MEHQ)
CAS Number: 177080-66-9
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 100
Specific Gravity (20/20): 1.13
Storage Temperature: 0°C


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: Methacrylic Acid 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Ester, 4-Methyl-2-oxotetrahydro-2H-pyran-4-yl Methacrylate

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Density: 1.13 (20 °C)
Flash Pt: 100 °C
CAS Number: 177080-66-9
https://us.vwr.com/stibo/web/std.lang.all/82/62/28988262.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR