6-Methoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 6-Methoxy-2-naphthylboronic acid6-Methoxy-2-naphthylboronic Acid

M2256-5G
TCM2256-5GEA 78.16 USD
TCM2256-5G
6-Methoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
6-Methoxy-2-naphthaleneboronic acid

CAS Number: 156641-98-4
MDL Number: MFCD03093087
Molecular Formula: C11H11BO3
Molecular Weight: 202.02
Form: Crystal
Color: White
Melting point (°C): 311


Other Product Names / Synonyms: 6-Methoxy-2-naphthylboronic Acid

Formula: C₁₁H₁₁BO₃
MW: 202.02 g/mol
Melting Pt: 311 °C
MDL Number: MFCD03093087
CAS Number: 156641-98-4
https://us.vwr.com/stibo/web/std.lang.all/82/50/28988250.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR