Molindone hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-1H-indol-4(5H)-one Hydrochloride3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one Hydrochloride

M2252-1G M2252-100MG
TCM2252-1GEA 284.82 USD
TCM2252-1G TCM2252-100MG
Molindone hydrochloride ≥98.0% (by HPLC, titration analysis)
Molindone hydrochloride

CAS Number: 15622-65-8
MDL Number: MFCD01718304
Molecular Formula: C16H24N2O2
Molecular Weight: 312.84
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 3-Ethyl-6,7-dihydro-2-methyl-5-(morpholinomethyl)-1H-indol-4(5H)-one Hydrochloride, 3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one Hydrochloride

Formula: C₁₆H₂₄N₂O₂
MW: 312.84 g/mol
MDL Number: MFCD01718304
CAS Number: 15622-65-8
https://us.vwr.com/stibo/web/std.lang.all/82/45/28988245.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR