3-(Trifluoromethyl)anisole ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Methoxy-3-(trifluoromethyl)benzene 3-Methoxybenzotrifluoride

M2243-5G M2243-25G
TCM2243-5GEA 39.57 USD
TCM2243-5G TCM2243-25G
3-(Trifluoromethyl)anisole ≥97.0% (by GC)
3-(Trifluoromethyl)anisole

CAS Number: 454-90-0
MDL Number: MFCD00000385
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 48
Specific Gravity (20/20): 1.24


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Methoxy-3-(trifluoromethyl)benzene, 3-(Trifluoromethyl)anisole

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 160 °C
Density: 1.24 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00000385
CAS Number: 454-90-0
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/82/39/28988239.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR