(Methylthio)acetaldehyde dimethyl acetal ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(Methylthio)acetaldehydedimethylacetal
CAS Number: 40015-15-4
MDL Number: MFCD00009848
Molecular Formula: C5H12O2S
Molecular Weight: 136.21
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): -19
Specific Gravity (20/20): 1.02
Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 1,1-Dimethoxy-2-(methylthio)ethane, (Methylmercapto)acetaldehyde Dimethyl Acetal
Formula:
C₅H₁₂O₂S MW: 136.21 g/mol Boiling Pt: 60 °C Density: 1.02 (20 °C) Flash Pt: –19 °C |
MDL Number:
MFCD00009848 CAS Number: 40015-15-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...