N-Methylpropylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-Methyl-1-propylamine

M2232-5G
TCM2232-5GEA 124.85 USD
TCM2232-5G
N-Methylpropylamine ≥98.0% (by GC)
N-Methylpropylamine

CAS Number: 627-35-0
MDL Number: MFCD00009361
Molecular Formula: C4H11N
Molecular Weight: 73.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): -32
Specific Gravity (20/20): 0.73


Other Product Names / Synonyms: N-Propylmethylamine

Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 63 °C
Density: 0.73 (20 °C)
Flash Pt: –32 °C
MDL Number: MFCD00009361
CAS Number: 627-35-0
https://us.vwr.com/stibo/web/std.lang.all/82/27/28988227.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR