2-Methylbenzhydryl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(alpha-Chlorobenzyl)tolueneChloro(2-methylphenyl)phenylmethane

M2211-25G M2211-5G
TCM2211-5GEA 73.05 USD
TCM2211-5G TCM2211-25G
2-Methylbenzhydryl chloride ≥98.0% (by GC, titration analysis)
2-Methylbenzhydryl chloride

CAS Number: 41870-52-4
MDL Number: MFCD00972765
Molecular Formula: C14H13Cl
Molecular Weight: 216.71
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 40
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane

Formula: C₁₄H₁₃Cl
MW: 216.71 g/mol
Boiling Pt: 136 °C
Melting Pt: 40 °C
MDL Number: MFCD00972765
CAS Number: 41870-52-4
https://us.vwr.com/stibo/web/std.lang.all/81/94/28988194.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR