(±)-4-Methylcyclohexene

Supplier: TCI America

Synonyms: 4-Methyl-1-cyclohexene

M2201-25G M2201-5G
TCM2201-5GEA 133.98 USD
TCM2201-5G TCM2201-25G
(±)-4-Methylcyclohexene
(±)-4-Methylcyclohexene

CAS Number: 591-47-9
MDL Number: MFCD00001573
Molecular Formula: C7H12
Molecular Weight: 96.17
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 103
Flash Point (°C): -1
Specific Gravity (20/20): 0.80

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 103 °C
Density: 0.80 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD00001573
CAS Number: 591-47-9
UN: 3295
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/81/85/28988185.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR