m-PEG12-NHS ester
Supplier: TCI America
Synonyms:
mPEG12-NHS Ester
M2188-25MG
TCM2188-25MGEA
244.19
USD
TCM2188-25MG
m-PEG12-NHS ester
m-PEG12-NHS ester
Molecular Weight: 685.76
Form: Paste
Color: White
Storage Temperature:
Molecular Formula: C30H55NO16
Other Product Names / Synonyms: N-Succinimidyl 4,7,10,13,16,19,22,25,28,31,34,37-Dodecaoxaoctatriacontanoate, 4,7,10,13,16,19,22,25,28,31,34,37-Dodecaoxaoctatriacontanoic Acid N-Succinimidyl Ester, mPEG12-NHS Ester
Formula:
C₃₀H₅₅NO₁₆ MW: 685.755 g/mol |
MDL Number:
MFCD13184970 CAS Number: 756525-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...