Methylmaleic hydrazide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Methyl-3,6-pyridazinediol3,6-Dihydroxy-4-methylpyridazine1,2-Dihydro-4-methyl-3,6-pyridazinedioneMethylmaleic Acid Hydrazide

M2182-25G M2182-5G
TCM2182-5GEA 96 USD
TCM2182-5G TCM2182-25G
Methylmaleic hydrazide ≥98.0% (by GC)
Methylmaleic hydrazide

CAS Number: 5754-18-7
MDL Number: MFCD00067746
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 287


Other Product Names / Synonyms: 1,2-Dihydro-4-methyl-3,6-pyridazinedione, 3,6-Dihydroxy-4-methylpyridazine, Methylmaleic Acid Hydrazide, 4-Methyl-3,6-pyridazinediol

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
Melting Pt: 287 °C
MDL Number: MFCD00067746
CAS Number: 5754-18-7
https://us.vwr.com/stibo/web/std.lang.all/81/66/28988166.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR