N-Methylmethanesulfonamide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: N-Methylmethane sulphonamideN-Methylmethane sulfonamide

M2170-25G
TCM2170-25GEA 58 USD
TCM2170-25G
N-Methylmethanesulfonamide ≥98.0% (by GC, total nitrogen)
N-Methylmethanesulfonamide

CAS Number: 1184-85-6
MDL Number: MFCD00776277
Molecular Formula: C2H7NO2S
Molecular Weight: 109.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Specific Gravity (20/20): 1.28

Formula: C₂H₇NO₂S
MW: 109.14 g/mol
Boiling Pt: 118 °C
Density: 1.28 (20 °C)
MDL Number: MFCD00776277
CAS Number: 1184-85-6
https://us.vwr.com/stibo/web/std.lang.all/81/50/28988150.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR