1-Methylpyrrole-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-Methyl-2-pyrrolecarboxylic acid
M2161-1G
TCM2161-1GEA
134
USD
TCM2161-1G
1-Methylpyrrole-2-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methylpyrrole-2-carboxylic acid
CAS Number: 6973-60-0
MDL Number: MFCD00003088
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 136
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Melting Pt: 136 °C |
MDL Number:
MFCD00003088 CAS Number: 6973-60-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...