5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 3-Cyano-5-methyl-2-(2-nitroanilino)thiophene5-Methyl-2-((2-nitrophenyl)amino)thiophene-3-carbonitrile

M2138-25G M2138-5G
TCM2138-5GEA 318.58 USD
TCM2138-5G TCM2138-25G
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile ≥97.0% (by GC)
5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile

CAS Number: 138564-59-7
MDL Number: MFCD06408054
Molecular Formula: C12H9N3O2S
Molecular Weight: 259.28
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 111


Other Product Names / Synonyms: 3-Cyano-5-methyl-2-(2-nitroanilino)thiophene

Formula: C₁₂H₉N₃O₂S
MW: 259.28 g/mol
Melting Pt: 111 °C
MDL Number: MFCD06408054
CAS Number: 138564-59-7
https://us.vwr.com/stibo/web/std.lang.all/81/04/28988104.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR