2-Methyl-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0% (by GC)
Supplier: TCI America
M2137-5G
TCM2137-5GEA
443
USD
TCM2137-5G
2-Methyl-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0% (by GC)
2-Methyl-4,6-bis(trichloromethyl)-1,3,5-triazine
CAS Number: 949-42-8
MDL Number: MFCD00023178
Molecular Formula: C6H3Cl6N3
Molecular Weight: 329.81
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 96
Formula:
C₆H₃Cl₆N₃ MW: 329.81 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00023178 CAS Number: 949-42-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...