1-(3-Methoxyphenyl)piperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2135-5G
M2135-25G
TCM2135-5GEA
60.9
USD
TCM2135-5G
TCM2135-25G
1-(3-Methoxyphenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(3-Methoxyphenyl)piperazine
CAS Number: 16015-71-7
MDL Number: MFCD00040733
Molecular Formula: C11H16N2O
Molecular Weight: 192.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 91
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: 3-(1-Piperazinyl)anisole
Formula:
C₁₁H₁₆N₂O MW: 192.26 g/mol Boiling Pt: 145 °C Density: 1.11 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00040733 CAS Number: 16015-71-7 UN: 3267 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...