(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: [(5-Sulphanyl-1,3,4-thiadiazol-2-yl)sulphanyl]acetic acid

M2100-25G M2100-5G
TCM2100-25GEA 425 USD
TCM2100-25G TCM2100-5G
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid ≥97.0% (by HPLC, titration analysis)
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid

CAS Number: 53723-88-9
MDL Number: MFCD00129970
Molecular Formula: C4H4N2O2S3
Molecular Weight: 208.27
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 167

Formula: C₄H₄N₂O₂S₃
MW: 208.28 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00129970
CAS Number: 53723-88-9
https://us.vwr.com/stibo/web/std.lang.all/80/55/28988055.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR