2-(Methylthio)benzaldehyde ≥97.0% (by GC)

Supplier: TCI America

M2094-1G
TCM2094-1GEA 145.15 USD
TCM2094-1G
2-(Methylthio)benzaldehyde ≥97.0% (by GC)
2-(Methylthio)benzaldehyde

CAS Number: 7022-45-9
MDL Number: MFCD00196822
Molecular Formula: C8H8OS
Molecular Weight: 152.21
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 149
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₈H₈OS
MW: 152.2135 g/mol
Boiling Pt: 149 °C
Flash Pt: 110 °C
MDL Number: MFCD00196822
CAS Number: 7022-45-9
https://us.vwr.com/stibo/web/std.lang.all/80/51/28988051.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR