2-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Methyl-3-pyridinecarboxylic acid
, 2-Methylpyridine-3-carboxylic acid
M2089-5G
M2089-1G
TCM2089-1GEA
91
USD
TCM2089-1G
TCM2089-5G
2-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
2-Methylnicotinic acid
CAS Number: 3222-56-8
MDL Number: MFCD00013449
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 227
Other Product Names / Synonyms: 2-Methylpyridine-3-carboxylic Acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Melting Pt: 227 °C |
MDL Number:
MFCD00013449 CAS Number: 3222-56-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...