1,1-Dimethylbiguanide hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Metformin hydrochloride

M2009-25G M2009-100G
TCM2009-25GEA 110.64 USD
TCM2009-25G TCM2009-100G
1,1-Dimethylbiguanide hydrochloride ≥98.0% (by HPLC, titration analysis)
1,1-Dimethylbiguanide hydrochloride

CAS Number: 1115-70-4
MDL Number: MFCD00012582
Molecular Formula: C4H11N5
Molecular Weight: 165.63
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 221
Lambda max.: 233 nm (H2O)


Product Categories: Bioscience, Obesity/Diabete Research Reagents, Regenerative Medicine Research Reagents, Antidiabetic Ingredients
Other Product Names / Synonyms: 1,1-Dimethylbiguanide Hydrochloride

Formula: C₄H₁₂ClN₅
MW: 165.625 g/mol
Melting Pt: 221 °C
MDL Number: MFCD00012582
CAS Number: 1115-70-4
https://us.vwr.com/stibo/web/std.lang.all/79/46/28987946.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR