2-Methyl-3-furanthiol ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 3-Mercapto-2-methylfuran

M1847-1G M1847-5G
TCM1847-5GEA 241.02 USD
TCM1847-5G TCM1847-1G
2-Methyl-3-furanthiol ≥95.0% (by GC)
2-Methyl-3-furanthiol

CAS Number: 28588-74-1
MDL Number: MFCD00010280
Molecular Formula: C5H6OS
Molecular Weight: 114.16
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 37
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C

Formula: C₅H₆OS
MW: 114.16 g/mol
Boiling Pt: 60 °C
Density: 1.12 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00010280
CAS Number: 28588-74-1
https://us.vwr.com/stibo/web/std.lang.all/62/61/28976261.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR