3-Mercaptobutanone ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
2-Mercapto-3-butanone
, 3-Mercapto-2-butanone
M1844-25G
TCM1844-25GEA
90.62
USD
TCM1844-25G
3-Mercaptobutanone ≥96.0% (by GC)
3-Mercaptobutanone
CAS Number: 40789-98-8
MDL Number: MFCD00085206
Molecular Formula: C4H8OS
Molecular Weight: 104.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 47
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C
Formula:
C₄H₈OS MW: 104.17 g/mol Boiling Pt: 90 °C Density: 1.03 (20 °C) Flash Pt: 47 °C |
MDL Number:
MFCD00085206 CAS Number: 40789-98-8 UN: 1224 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...