2,6-Dinitro-4-methylanisole ≥98.0% (by GC)
Supplier: TCI America
M1842-5G
TCM1842-5GEA
157
USD
TCM1842-5G
2,6-Dinitro-4-methylanisole ≥98.0% (by GC)
2,6-Dinitro-4-methylanisole
CAS Number: 29455-11-6
MDL Number: MFCD00192097
Molecular Formula: C8H8N2O5
Molecular Weight: 212.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 124
Other Product Names / Synonyms: 4-Methyl-2,6-dinitroanisole, 1-Methoxy-4-methyl-2,6-dinitrobenzene
Formula:
C₈H₈N₂O₅ Melting Pt: 124 °C |
MDL Number:
MFCD00192097 CAS Number: 29455-11-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...