2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Mercapto-2-methyltetrahydrofuran2-Methyl-3-tetrahydrofuranthiol

M1841-1G M1841-5G
TCM1841-1GEA 28.84 USD
TCM1841-1G TCM1841-5G
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) ≥98.0% (by GC)
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture)

CAS Number: 57124-87-5
MDL Number: MFCD01632596
Molecular Formula: C5H10OS
Molecular Weight: 118.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 51
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 3-Mercapto-2-methyltetrahydrofuran

Formula: C₅H₁₀OS
MW: 118.19 g/mol
Boiling Pt: 168 °C
Density: 1.05 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD01632596
CAS Number: 57124-87-5
https://us.vwr.com/stibo/web/std.lang.all/62/49/28976249.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR