α-Chloro-2-methyl-3-nitrotoluene ≥98.0% (by GC)
Supplier: TCI America
M1816-5G
TCM1816-5GEA
71
USD
TCM1816-5G
α-Chloro-2-methyl-3-nitrotoluene ≥98.0% (by GC)
α-Chloro-2-methyl-3-nitrotoluene
CAS Number: 60468-54-4
MDL Number: MFCD00007165
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 113
Other Product Names / Synonyms: 2-(Chloromethyl)-6-nitrotoluene
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol Melting Pt: 45 °C Flash Pt: 113 °C |
MDL Number:
MFCD00007165 CAS Number: 60468-54-4 UN: 3261 ADR: 8,II |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...