(S)-(-)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)

Synonyms: (S)-(-)-beta-Methylphenethylamine(S)-(-)-2-Phenyl-1-propylamine

M1747-5G M1747-25G
TCM1747-5GEA 147.63 USD
TCM1747-5G TCM1747-25G
(S)-(-)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-β-Methylphenethylamine

CAS Number: 17596-79-1
MDL Number: MFCD00216741
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 81
Specific Gravity (20/20): 0.95
Specific rotation [a]20/D: -32 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Phenyl-1-propylamine

Formula: C₉H₁₃N
Density: 0.95 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00216741
CAS Number: 17596-79-1
https://us.vwr.com/stibo/web/std.lang.all/60/95/28976095.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR