Gal[246Bn]β(1-4)Glc[236Bn]-β-MP ≥95.0% (by HPLC)

Synonyms: LacMP 3'-OHper OBn4-Methoxyphenyl 4-O-(2,4,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside

M1727-1G
TCM1727-1GEA 420.24 USD
TCM1727-1G
Gal[246Bn]β(1-4)Glc[236Bn]-β-MP ≥95.0% (by HPLC)
Gal[246Bn]beta(1-4)Glc[236Bn]-beta-MP

CAS Number: 717132-49-5
Molecular Formula: C61H64O12
Molecular Weight: 989.17
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -8 deg (C=0.4, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Glycosylation, Glycosylation, Bioscience, Disaccharides, Glycosides, p-Methoxyphenyl Glycosides, O-Substituted Sugars
Other Product Names / Synonyms: LacMP 3'-OH, per OBn, 4-Methoxyphenyl 4-O-(2,4,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside

Formula: C₆₁H₆₄O₁₂
MW: 989.17 g/mol
CAS Number: 717132-49-5
https://us.vwr.com/stibo/web/std.lang.all/60/56/28976056.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR