2-Methoxy-6-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1695-5G M1695-25G
TCM1695-25GEA 87 USD
TCM1695-25G TCM1695-5G
2-Methoxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Methoxy-6-methylpyridine

CAS Number: 63071-03-4
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 156
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 6-Methoxy-2-picoline

Formula: C₇H₉NO
MW: 123.16 g/mol
Boiling Pt: 156 °C
Density: 1.01 (20 °C)
MDL Number: MFCD04112507
CAS Number: 63071-03-4
https://us.vwr.com/stibo/web/std.lang.all/60/06/28976006.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR